(S)-(+)-1-Benzyl-3-hydroxypiperidine 97%

Warning

H30615-MD H30615-03
AAH30615-MDEA 75.06 USD
AAH30615-MD AAH30615-03
(S)-(+)-1-Benzyl-3-hydroxypiperidine 97%
(S)-(+)-1-Benzyl-3-hydroxypiperidine
Formula: C₁₂H₁₇NO
Storage Temperature: Ambient
MDL Number: MFCD03426359
CAS Number: 91599-79-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR