(S)-(+)-3-Amino-1-boc-piperidine 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: (S)-1-Boc-3-aminopiperidine(S)-(+)-3-Amino-1-BOC-piperidine(S)-tert-Butyl 3-aminopiperidine-1-carboxylate

H30538-03 H30538-06 H30538-MD
AAH30538-MDEA 85.13 USD
AAH30538-MD AAH30538-06 AAH30538-03
(S)-(+)-3-Amino-1-boc-piperidine 97%
(S)-(+)-3-Amino-1-Boc-piperidine
Formula: C₁₀H₂₀N₂O₂
MW: 200.28 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03094718
CAS Number: 625471-18-3
UN: 2735
ADR: "8",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR