2-Amino-5-chlorobenzyl alcohol 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (2-Amino-5-chlorophenyl)methanol

H30531-06 H30531-14
AAH30531-14EA 307.99 USD
AAH30531-14 AAH30531-06
2-Amino-5-chlorobenzyl alcohol 98%
2-Amino-5-chlorobenzyl alcohol
Formula: C₇H₈ClNO
MW: 157.60 g/mol
Melting Pt: 107…109 °C
Storage Temperature: Ambient
MDL Number: MFCD00075163
CAS Number: 37585-25-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR