2-(4-Chlorophenyl)ethylboronic acid pinacol ester 98%

Supplier: Thermo Scientific Chemicals

Synonyms: 2-(4-Chlorophenethyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

H30303-06 H30303-03
AAH30303-06EA 120 USD
AAH30303-06 AAH30303-03
2-(4-Chlorophenyl)ethylboronic acid pinacol ester 98%
2-(4-Chlorophenyl)ethylboronic acid pinacol ester
Formula: C₁₄H₂₀BClO₂
MW: 266.58 g/mol
Melting Pt: 34…38 °C
Storage Temperature: Ambient
MDL Number: MFCD12546184
CAS Number: 444094-88-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR