5-(Trifluoromethoxy)-m-toluidine 97%, JRD

Warning

H30206-MD H30206-03
AAH30206-03EA 306.36 USD
AAH30206-03 AAH30206-MD
5-(Trifluoromethoxy)-m-toluidine 97%, JRD
5-(Trifluoromethoxy)-m-toluidine
Formula: C₈H₈F₃NO
MW: 191.15 g/mol
Boiling Pt: 73 °C/1 mm
Storage Temperature: Ambient
MDL Number: MFCD09025380
CAS Number: 86256-63-5
UN: 2810
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR