2-Aminothiazole-5-carbonitrile 98%

Warning

H30019-03
AAH30019-03EA 109.77 USD
AAH30019-03
2-Aminothiazole-5-carbonitrile 98%
2-Aminothiazole-5-carbonitrile
Formula: C₄H₃N₃S
MW: 125.20 g/mol
Melting Pt: ca 185 °C
Storage Temperature: Ambient
MDL Number: MFCD08275724
CAS Number: 51640-52-9
UN: 3439
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR