3-(4-Methyl-1-piperazinylcarbonyl)phenylboronic acid pinacol ester 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
(4-Methylpiperazin-1-yl)(3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)methanone
, 3-(4-Methyl-1-piperazinylcarbonyl)benzeneboronic acid pinacol ester
H29224-06
H29224-03
AAH29224-06EA
290.99
USD
AAH29224-06
AAH29224-03
3-(4-Methyl-1-piperazinylcarbonyl)phenylboronic acid pinacol ester 97%
3-(4-Methyl-1-piperazinylcarbonyl)phenylboronic acid pinacol ester
Formula:
C₁₈H₂₇BN₂O₃ MW: 330.23 g/mol Melting Pt: 86…89 °C Storage Temperature: Ambient |
MDL Number:
MFCD05864308 CAS Number: 883738-38-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...