4-Bromo-2-nitrophenylhydrazine hydrochloride 95%

Supplier: Thermo Scientific Chemicals
Warning

H29202-03 H29202-06
AAH29202-03EA 126.99 USD
AAH29202-03 AAH29202-06
4-Bromo-2-nitrophenylhydrazine hydrochloride 95%
4-Bromo-2-nitrophenylhydrazine hydrochloride
Formula: C₆H₇BrClN₃O₂
Melting Pt: ca 185 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD09260482
CAS Number: 100032-79-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR