4-Chlorothieno[3.2.c]pyridine 98%

Warning

H29157-ME
AAH29157-MEEA 287.49 USD
AAH29157-ME
4-Chlorothieno[3.2.c]pyridine 98%
4-Chlorothieno[3.2.c]pyridine
Formula: C₇H₄ClNS
MW: 169.63 g/mol
Melting Pt: 99…101 °C
Storage Temperature: Ambient
MDL Number: MFCD01861830
CAS Number: 27685-94-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR