(R)-(+)-1,2,3,4-Tetrahydroisoquinoline-3-carboxylic acid
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
H-D-Tic-OH
, (R)-1,2,3,4-Tetrahydro-3-isoquinolinecarboxylic Acid
, D-1,2,3,4-Tetrahydroisoquinoline-3-carboxylic acid
H29112-MD
H29112-06
H29112-03
AAH29112-03EA
247.99
USD
AAH29112-03
AAH29112-MD
AAH29112-06
(R)-(+)-1,2,3,4-Tetrahydroisoquinoline-3-carboxylic acid
(R)-(+)-1,2,3,4-Tetrahydroisoquinoline-3-carboxylic acid
Formula:
C₁₀H₁₁NO₂ MW: 177.2 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00144038 CAS Number: 103733-65-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...