Cyclopropanesulfonyl chloride 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: CyclopropanesulphonylchlorideCyclopropanesulfonylchloride

H29107-03 H29107-06
AAH29107-06EA 170.99 USD
AAH29107-06 AAH29107-03
Cyclopropanesulfonyl chloride 97%
Cyclopropanesulphonyl chloride
Formula: C₃H₅ClO₂S
MW: 140.59 g/mol
Boiling Pt: 60 °C/2 mm
Density: 1.39
Flash Pt: 96 °C(205 °F)
Storage Temperature: Ambient
MDL Number: MFCD01631933
CAS Number: 139631-62-2
UN: 3265
ADR: "8",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR