2,2-Difluorocyclopropylamine hydrochloride 97%
|
Warning
|
H29051-MC
H29051-ME
AAH29051-MEEA
628.3
USD
AAH29051-ME
AAH29051-MC
2,2-Difluorocyclopropylamine hydrochloride 97%
2,2-Difluorocyclopropylamine hydrochloride
Formula:
C₃H₆ClF₂N MW: 129.54 g/mol Melting Pt: ca 145 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD07777150 CAS Number: 105614-25-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...