n-Dodecanethiol gold nanoparticles 2% (w/v) in toluene 2-5nm particles

Supplier: Alfa Aesar
Danger

H29045-AA
AAH29045-AAEA 23.83 USD
AAH29045-AA
n-Dodecanethiol gold nanoparticles 2% (w/v) in toluene 2-5nm particles
n-Dodecanethiol gold nanoparticles
Storage Temperature: Ambient
MDL Number: MFCD09953506
UN: 1294
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR