4-Amino-1H-pyrazole 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1H-Pyrazol-4-amine4-Aminopyrazole

H28921-ME H28921-MC
AAH28921-MCEA 75.19 USD
AAH28921-MC AAH28921-ME
4-Amino-1H-pyrazole 95%
4-Amino-1H-pyrazole
Formula: C₃H₅N₃
MW: 83.09 g/mol
Melting Pt: 72…77 °C
Storage Temperature: Ambient
MDL Number: MFCD01693729
CAS Number: 28466-26-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR