3-Methoxybenzenesulfonamide 97%

Supplier: Thermo Scientific Chemicals

Warning

H28844-03
AAH28844-03EA 99.6 USD
AAH28844-03
3-Methoxybenzenesulfonamide 97%
3-Methoxybenzenesulphonamide
Formula: C₇H₉NO₃S
Melting Pt: 131…133 °C
Storage Temperature: Ambient
MDL Number: MFCD08704579
CAS Number: 58734-57-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR