3-(1-Pyrrolidinylcarbonyl)phenylboronic acid pinacol ester 97%

Synonyms: 3-(1-Pyrrolidinylcarbonyl)benzeneboronic acid pinacol ester

H28801-03
AAH28801-03EA 61.92 USD
AAH28801-03
3-(1-Pyrrolidinylcarbonyl)phenylboronic acid pinacol ester 97%
3-(1-Pyrrolidinylcarbonyl)phenylboronic acid pinacol ester
Formula: C₁₇H₂₄BNO₃
MW: 301.19
Melting Pt: 96…98 °C
Storage Temperature: Ambient
MDL Number: MFCD09027086
CAS Number: 1073353-61-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR