4-Methyl-2-(methylthio)pyrimidine 98%

Supplier: Thermo Scientific Chemicals

Warning

Synonyms: 4-Methyl-2(methylsulfanyl)pyrimidine

H28795-06
AAH28795-06EA 66.5 USD
AAH28795-06
4-Methyl-2-(methylthio)pyrimidine 98%
4-Methyl-2-(methylthio)pyrimidine
Formula: C₆H₈N₂S
MW: 140.21 g/mol
Density: 1.01
Flash Pt: 104 °C(219 °F)
Storage Temperature: Ambient
MDL Number: MFCD00023242
CAS Number: 14001-63-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR