7-Benzyloxyindole-3-carbaldehyde 97%

Warning

H28777-MD
AAH28777-MDEA 67.73 USD
AAH28777-MD
7-Benzyloxyindole-3-carbaldehyde 97%
7-Benzyloxyindole-3-carbaldehyde
Formula: C₁₆H₁₃NO₂
MW: 251.28 g/mol
Melting Pt: 152…159 °C
Storage Temperature: Ambient
MDL Number: MFCD00055979
CAS Number: 92855-65-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR