2-Amino-3,5-dibromopyrazine 99%
|
Warning
|
H28744-03
AAH28744-03EA
90.43
USD
AAH28744-03
2-Amino-3,5-dibromopyrazine 99%
2-Amino-3,5-dibromopyrazine
Formula:
C₄H₃Br₂N₃ MW: 252.90 g/mol Melting Pt: 114…117 °C Storage Temperature: Ambient |
MDL Number:
MFCD00673150 CAS Number: 24241-18-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...