1-(p-Toluenesulfonyl)pyrrole 98%
|
Warning
|
Synonyms:
1-Tosylpyrrole
, 1-Tosyl-1H-pyrrole
, N-Tosylpyrrole
H28641-06
H28641-14
AAH28641-06EA
38.12
USD
AAH28641-06
AAH28641-14
1-(p-Toluenesulfonyl)pyrrole 98%
1-(p-Toluenesulfonyl)pyrrole
Formula:
C₁₁H₁₁NO₂S MW: 221.27 g/mol Melting Pt: 99…103 °C Storage Temperature: Ambient |
MDL Number:
MFCD00145014 CAS Number: 17639-64-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...