3-(Furfurylaminocarbonyl)benzeneboronic acid pinacol ester 97%

H28599-03 H28599-06
AAH28599-06EA 293.2 USD
AAH28599-06 AAH28599-03
3-(Furfurylaminocarbonyl)benzeneboronic acid pinacol ester 97%
3-(Furfurylaminocarbonyl)benzeneboronic acid pinacol ester
Formula: C₁₈H₂₂BNO₄
MW: 327.18 g/mol
Melting Pt: 122…124 °C
Storage Temperature: Ambient
MDL Number: MFCD09266173
CAS Number: 1073353-63-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR