(±)-3-Amino-1-(benzyloxycarbonyl)pyrrolidine
|
Warning
|
Synonyms:
Benzyl-3-aminopyrrolidine-1-carboxylate
, 1-Benzyloxycarbonyl-3-aminopyrrolidine
, 1-Z-3-aminopyrrolidine
, 1-Cbz-3-aminopyrrolidine
H28525-03
H28525-MD
AAH28525-MDEA
54.46
USD
AAH28525-MD
AAH28525-03
(±)-3-Amino-1-(benzyloxycarbonyl)pyrrolidine
(±)-3-Amino-1-(benzyloxycarbonyl)pyrrolidine
-
Formula:
C₁₂H₁₆N₂O₂ MW: 220.27 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD03001673 CAS Number: 185057-50-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...