Fmoc-D-Pro-OH, (max. 5% H₂O) 98+%

Supplier: Thermo Scientific Chemicals

Synonyms: N-Fmoc-D-proline

H28500-06 H28500-03 H28500-14
AAH28500-03EA 34.39 USD
AAH28500-03 AAH28500-14 AAH28500-06
Fmoc-D-Pro-OH, (max. 5% H₂O) 98+%
Fmoc-D-Pro-OH
Formula: C₂₀H₁₉NO₄
MW: 337.38 g/mol
Melting Pt: 110…116 °C
Storage Temperature: Ambient
MDL Number: MFCD00077067
CAS Number: 101555-62-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR