4-Chloro-6,7-dimethoxyquinazoline 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H28405-14
H28405-06
AAH28405-14EA
231.99
USD
AAH28405-14
AAH28405-06
4-Chloro-6,7-dimethoxyquinazoline 98%
4-Chloro-6,7-dimethoxyquinazoline
Formula:
C₁₀H₉ClN₂O₂ MW: 224.65 g/mol Melting Pt: 184…187 °C Storage Temperature: Ambient |
MDL Number:
MFCD01570172 CAS Number: 13790-39-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...