Methyl thiosalicylate 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Methyl 2-mercaptobenzoate

H28400-06 H28400-14
AAH28400-06EA 94.39 USD
AAH28400-06 AAH28400-14
Methyl thiosalicylate 97%
Methyl thiosalicylate
Formula: C₈H₈O₂S
MW: 168.21 g/mol
Boiling Pt: 98…100 °C/2 mm
Density: 1.223
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00060678
CAS Number: 4892-02-8
UN: 2810
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR