1,2-Dimethoxy-1,1,2,2-tetramethyldisilane 97%

Warning

H28370-03 H28370-MD H28370-06
AAH28370-03EA 102.34 USD
AAH28370-03 AAH28370-MD AAH28370-06
1,2-Dimethoxy-1,1,2,2-tetramethyldisilane 97%
1,2-Dimethoxy-1,1,2,2-tetramethyldisilane
Formula: C₆H₁₈Si₂O₂
Density: 0.859
Flash Pt: 27 °C(81 °F)
Storage Temperature: Ambient
MDL Number: MFCD09038446
CAS Number: 10124-62-6
UN: 1993
ADR: "3",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR