2-Methylphenethylamine 98%
|
Danger
|
Synonyms:
2-(o-Tolyl)ethylamine
H28320-06
AAH28320-06EA
163.8
USD
AAH28320-06
2-Methylphenethylamine 98%
2-Methylphenethylamine
Formula:
C₉H₁₃N Density: 0.96 Storage Temperature: Ambient |
MDL Number:
MFCD01310827 CAS Number: 55755-16-3 UN: 2735 ADR: "8",III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...