3-Carboxy-4-fluorophenylboronic acid 97%

Warning

Synonyms: 3-Carboxy-4-fluorophenylboronic Acid3-Carboxy-4-fluorobenzeneboronic acid

H28313-09 H28313-03
AAH28313-09EA 154.29 USD
AAH28313-09 AAH28313-03
3-Carboxy-4-fluorophenylboronic acid 97%
3-Carboxy-4-fluorophenylboronic acid
Formula: C₇H₆BFO₄
MW: 183.93 g/mol
Melting Pt: 217…219 °C
Storage Temperature: Ambient
MDL Number: MFCD03095129
CAS Number: 872460-12-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR