5-Phenyl-o-toluidine 98%

Supplier: Alfa Aesar
Warning

Synonyms: 3-Amino-4-methylbiphenyl

H28265-03
AAH28265-03EA 52.75 USD
AAH28265-03
5-Phenyl-o-toluidine 98%
5-Phenyl-o-toluidine
Formula: C₁₃H₁₃N
Storage Temperature: Ambient
CAS Number: 80938-67-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR