4-tert-Butylthiophenol gold 2% (w/v) in toluene, nanoparticles 2 -5 nm

Danger

H28235-AA
AAH28235-AAEA 399.6 USD
AAH28235-AA
4-tert-Butylthiophenol gold 2% (w/v) in toluene, nanoparticles 2 -5 nm
4-tert-Butylthiophenol gold in toluene
Boiling Pt: 111 °C
Flash Pt: 4 °C(39 °F)
Storage Temperature: Ambient
MDL Number: MFCD09881474
UN: 1294
ADR: "3",II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR