4-(4-Methyl-1-piperazinylcarbonyl)phenylboronic acid pinacol ester 97%

Warning

Synonyms: (4-Methylpiperazin-1-yl)(4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)methanone4-(4-Methyl-1-piperazinylcarbonyl)benzeneboronic acid pinacol ester

H28216-03 H28216-06
AAH28216-06EA 200.53 USD
AAH28216-06 AAH28216-03
4-(4-Methyl-1-piperazinylcarbonyl)phenylboronic acid pinacol ester 97%
4-(4-Methyl-1-piperazinylcarbonyl)phenylboronic acid pinacol ester
Formula: C₁₈H₂₇BN₂O₃
MW: 330.23 g/mol
Melting Pt: 110…113 °C
Storage Temperature: Ambient
MDL Number: MFCD05863914
CAS Number: 832114-06-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR