(3S,4R)-4-(4-Fluorophenyl)-3-hydroxymethyl-1-methylpiperidine 98%

Danger

H28137-14 H28137-06
AAH28137-14EA 188.7 USD
AAH28137-14 AAH28137-06
(3S,4R)-4-(4-Fluorophenyl)-3-hydroxymethyl-1-methylpiperidine 98%
(3S,4R)-4-(4-Fluorophenyl)-3-hydroxymethyl-1-methylpiperidine
Formula: C₁₃H₁₈FNO
MW: 223.29 g/mol
Melting Pt: 93…98 °C
Storage Temperature: Ambient
MDL Number: MFCD06658161
CAS Number: 105812-81-5
EINECS: 406-030-4
UN: 2811
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR