Potassium-3-formylphenyltrifluoroborate 97%
|
Warning
|
Synonyms:
Potassium trifluoro(3-formylphenyl)borate
H28125-06
H28125-03
AAH28125-06EA
108.15
USD
AAH28125-06
AAH28125-03
Potassium-3-formylphenyltrifluoroborate 97%
Potassium 3-formylphenyltrifluoroborate
Formula:
C₇H₅BF₃KO MW: 212.02 g/mol Melting Pt: ca 200 °C Storage Temperature: Ambient |
MDL Number:
MFCD07784378 CAS Number: 871231-44-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...