Ethyl-α-4-dibromophenylacetate ≥97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Ethyl 2-bromo-2-(4-bromophenyl)acetate
H28093-06
H28093-14
AAH28093-14EA
483.99
USD
AAH28093-14
AAH28093-06
Ethyl-α-4-dibromophenylacetate ≥97%
Ethyl-alpha-4-dibromophenylacetate
Formula:
C₁₀H₁₀Br₂O₂ MW: 321.99 g/mol Boiling Pt: 140 °C/5 mm Density: 1.642 Flash Pt: >110 °C(230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD09953497 CAS Number: 77143-76-1 UN: 3265 ADR: "8",III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...