3-Benzoyl-2-thiophenecarboxylic acid 96%

Warning

H28040-MC H28040-MD H28040-03
AAH28040-03EA 410.7 USD
AAH28040-03 AAH28040-MC AAH28040-MD
3-Benzoyl-2-thiophenecarboxylic acid 96%
3-Benzoyl-2-thiophenecarboxylic acid
Formula: C₁₂H₈O₃S
Melting Pt: 150…153 °C
Storage Temperature: Ambient
MDL Number: MFCD00159542
CAS Number: 30006-03-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR