2-Ethoxybenzonitrile 98%

Warning

H28012-03
AAH28012-03EA 43.01 USD
AAH28012-03
2-Ethoxybenzonitrile 98%
2-Ethoxybenzonitrile
Formula: C₉H₉NO
MW: 147.18 g/mol
Density: 1.053
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001789
CAS Number: 6609-57-0
UN: 3276
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR