3,3-Dimethylcyclohexanone 97%

Supplier: Thermo Scientific Chemicals
Warning

H28000-03 H28000-06
AAH28000-06EA 97.49 USD
AAH28000-06 AAH28000-03
3,3-Dimethylcyclohexanone 97%
3,3-Dimethylcyclohexanone
Formula: C₈H₁₄O
MW: 126.2 g/mol
Boiling Pt: 175 °C
Melting Pt: 6 °C
Density: 0.91
Flash Pt: 59 °C(138 °F)
Storage Temperature: Ambient
MDL Number: MFCD00040178
CAS Number: 2979-19-3
UN: 1224
ADR: "3",III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR