(2S,3aS,7aS)-2-Carboxyoctahydroindole 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
(2S,3aS,7aS)-Octahydro-1H-indole-2-carboxylic acid
, H-Oic-OH
H27894-06
H27894-03
AAH27894-03EA
77.09
USD
AAH27894-03
AAH27894-06
(2S,3aS,7aS)-2-Carboxyoctahydroindole 98%
(2S,3aS,7aS)-2-Carboxyoctahydroindole
Formula:
C₉H₁₅NO₂ MW: 169.22 g/mol Melting Pt: 266 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD07782125 CAS Number: 80875-98-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...