5-Bromo-2,3'-bipyridyl 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 5-Bromo-2,3'-bipyridine

H27880-03
AAH27880-03EA 326.99 USD
AAH27880-03
5-Bromo-2,3'-bipyridyl 95%
5-Bromo-2,3'-bipyridyl
Formula: C₁₀H₇BrN₂
MW: 235.08
Melting Pt: 75…78 °C
Storage Temperature: Ambient
MDL Number: MFCD09037420
CAS Number: 774-53-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR