Methyl thiocyanate 99%

Supplier: Thermo Scientific Chemicals
Danger

H27879-AC H27879-AE H27879-AA
AAH27879-AAEA 31.69 USD
AAH27879-AA AAH27879-AC AAH27879-AE
Methyl thiocyanate 99%
Methyl thiocyanate
Formula: C₂H₃NS
MW: 73.12 g/mol
Boiling Pt: 131 °C
Melting Pt: –5 °C
Density: 1.068
Flash Pt: 38 °C(100 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001830
CAS Number: 556-64-9
EINECS: 209-134-6
UN: 1992
ADR: "3",III
Merck Index: 12,06209

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR