3-Cyanocoumarin 97%

Synonyms: Coumarin-3-carbonitrile2-Oxo-2H-chromene-3-carbonitrile

H27871-03 H27871-06
AAH27871-06EA 101.36 USD
AAH27871-06 AAH27871-03
3-Cyanocoumarin 97%
3-Cyanocoumarin
Formula: C₁₀H₅NO₂
MW: 171.16 g/mol
Melting Pt: 184…188 °C
Storage Temperature: Ambient
MDL Number: MFCD00115699
CAS Number: 15119-34-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR