(1S,2S)-N-Methylsulfonyl-1,2-diphenylethanediamine ≥98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
N-((1S,2S)-2-Amino-1,2-diphenylethyl)methanesulfonamide
, N-((1S,2S)-2-Amino-1,2-diphenylethyl)methanesulphonamide
H27867-14
H27867-03
H27867-06
AAH27867-14EA
1508.49
USD
AAH27867-14
AAH27867-03
AAH27867-06
(1S,2S)-N-Methylsulfonyl-1,2-diphenylethanediamine ≥98%
(1S,2S)-N-Methylsulfonyl-1,2-diphenylethanediamine
Formula:
C₁₅H₁₈N₂O₂S MW: 290.38 g/mol Melting Pt: 117…120 °C Storage Temperature: Ambient |
MDL Number:
MFCD03791037 CAS Number: 300345-76-0 UN: 3259 ADR: "8",III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...