2,5-Dichloro-3-pyridineboronic acid 95%

Warning

Synonyms: 2,5-Dichloropyridine-3-boronic acid

H27849-03
AAH27849-03EA 329.67 USD
AAH27849-03
2,5-Dichloro-3-pyridineboronic acid 95%
2,5-Dichloro-3-pyridineboronic acid
Formula: C₅H₄BCl₂NO₂
MW: 191.81 g/mol
Melting Pt: 152…155 °C
Storage Temperature: Ambient
MDL Number: MFCD06659486
CAS Number: 536693-97-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR