3,5-Difluoro-L-phenylalanine 98%

Synonyms: (S)-2-Amino-3-(3,5-difluorophenyl)propanoic acid

H27833-MD H27833-03
AAH27833-MDEA 105.56 USD
AAH27833-MD AAH27833-03
3,5-Difluoro-L-phenylalanine 98%
3,5-Difluoro-L-phenylalanine
Formula: C₉H₉F₂NO₂
MW: 201.18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01631987
CAS Number: 31105-91-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR