Methyl (+/-)-1-benzyloxycarbonyl-4-boc-piperazine-2-carboxylate
|
Warning
|
H27832-03
AAH27832-03EA
149.85
USD
AAH27832-03
Methyl (+/-)-1-benzyloxycarbonyl-4-boc-piperazine-2-carboxylate
Methyl (+/-)-1-benzyloxycarbonyl-4-boc-piperazine-2-carboxylate
Formula:
C₁₉H₂₆N₂O₆ MW: 378.43 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD03426296 CAS Number: 126937-42-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...