3-Cyano-6-methylchromone 99%

Warning

Synonyms: 6-Methyl-4-oxo-4H-chromene-3-carbonitrile6-Methylchromone-3-carbonitrile

H27825-03 H27825-06
AAH27825-03EA 45.32 USD
AAH27825-03 AAH27825-06
3-Cyano-6-methylchromone 99%
3-Cyano-6-methylchromone
Formula: C₁₁H₇NO₂
MW: 185.19 g/mol
Melting Pt: 150…152 °C
Storage Temperature: Ambient
MDL Number: MFCD00191962
CAS Number: 50743-18-5
UN: 3439
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR