2-Chloro-5-pentylpyrimidine 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H27808-09
H27808-03
AAH27808-09EA
176.99
USD
AAH27808-09
AAH27808-03
2-Chloro-5-pentylpyrimidine 98%
2-Chloro-5-pentylpyrimidine
Formula:
C₉H₁₃ClN₂ Boiling Pt: 124 °C/3 mm Density: 1.085 Flash Pt: 97 °C(207 °F) Storage Temperature: Ambient |
MDL Number:
MFCD04038093 CAS Number: 154466-62-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...