2-Fluoro-L-phenylalanine 99+% (sum of enantiomers), Powder
Synonyms:
H-Phe(2-F)-OH
H27807-MD
H27807-03
AAH27807-MDEA
69.07
USD
AAH27807-MD
AAH27807-03
2-Fluoro-L-phenylalanine 99+% (sum of enantiomers), Powder
2-Fluoro-L-phenylalanine
Formula:
C₉H₁₀FNO₂ MW: 183.2 g/mol Storage Temperature: Ambient |
CAS Number:
19883-78-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...