3-Bromo-2,6-dichloropyridine 95%
|
Warning
|
H27797-03
H27797-06
AAH27797-06EA
779.22
USD
AAH27797-06
AAH27797-03
3-Bromo-2,6-dichloropyridine 95%
3-Bromo-2,6-dichloropyridine
Formula:
C₅H₂BrCl₂N Melting Pt: 68…71 °C Storage Temperature: Ambient |
MDL Number:
MFCD06798231 CAS Number: 866755-20-6 UN: 2811 ADR: "6.1",III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...