3-Chloro-2-fluoro-4-pyridineboronic acid 95%
|
Warning
|
Synonyms:
(3-Chloro-2-fluoropyridin-4-yl)boronic acid
H27781-MD
H27781-03
AAH27781-03EA
334.11
USD
AAH27781-03
AAH27781-MD
3-Chloro-2-fluoro-4-pyridineboronic acid 95%
3-Chloro-2-fluoro-4-pyridineboronic acid
Formula:
C₅H₄BClFNO₂ MW: 175.35 g/mol Melting Pt: 126…130 °C Storage Temperature: Ambient |
MDL Number:
MFCD09037477 CAS Number: 1217500-55-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...